Provette per centrifuga
Supplier: DELTALAB
Jump to:
About this item
Tubes made of PP, translucent, with blue HDPE screw cap.
- Continuous thread
- External moulded graduations in 0,5 ml increments
- Excellent temperature resistance
- Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.
9 Options Available Below
Product Details & Documents
Tubes made of PP, translucent, with blue HDPE screw cap.
- Continuous thread
- External moulded graduations in 0,5 ml increments
- Excellent temperature resistance
- Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.
Product Family Options
Centrifuge tubes 15 ml
Product Information
- DescrizioneCapacitàØ×LSteriliTipo di chiusuraRCF max.TipoVersioneConfezioneAvailabilityPrice
- Centrifuge tubes, non-sterile15 ml17×122 mmTappo a vite7000×gWithout skirtNon sterileBulkLog in for availability1 * 500 STSFr. 155.00
- 15 ml17×122 mmTappo a vite5000×gWithout skirtSterile100 per bag/500 per caseLog in for availability1 * 500 STSFr. 254.00
- 15 ml17×122 mmTappo a vite5000×gWithout skirtSterileIndividually baggedLog in for availability1 * 500 STSFr. 253.00
Centrifuge tubes 50 ml
Product Information
- DescrizioneCapacitàØ×LSteriliTipo di chiusuraRCF max.TipoVersioneConfezioneAvailabilityPrice
- 50 ml29×118,5 mmTappo a vite12000×gSenza bordoNon sterileBulkLog in for availability1 * 500 STSFr. 200.00
- Centrifuge tubes, skirted, non-sterile50 ml29×118,5 mm-12000×gCon bordoNon sterileBulkLog in for availability1 * 500 STSFr. 214.00
- 50 ml29×118,5 mmTappo a vite7000×gSenza bordoSterileIndividually baggedLog in for availability1 * 500 STSFr. 343.00
- Centrifuge tubes, skirted, sterile50 ml29×118,5 mm+Tappo a vite7000×gCon bordoSterileIndividually baggedLog in for availability1 * 500 STSFr. 382.00
- 50 ml29×118,5 mmTappo a vite7000×gSkirtedSterile100 per bag/500 per caseLog in for availability1 * 500 STSFr. 377.00
- 50 ml29×118,5 mmTappo a vite7000×gSkirtedSterile25 per bag/500 per caseLog in for availability1 * 500 STSFr. 393.00