XTRA™ Carts
Informazioni su questo articolo
Durable and attractive storage and transportation carts for front and back-of house applications.
- Metal uprights and moulded shelves engineered for durability and appearance
- Non-marking 10,2 cm swivel casters
- Wide shelves hold a variety of equipment and supplies, up to 135 kg
- Available in various versions
Accessories information: Accessories are available for a complete system solution for housekeeping in the hospitality industry. Utility bin features a smooth easy-to-clean surface.
Informazioni per l'ordine: Attachable plastic container must be ordered separately as an accessory.
5 varianti
Product Details & Documents
Durable and attractive storage and transportation carts for front and back-of house applications.
- Metal uprights and moulded shelves engineered for durability and appearance
- Non-marking 10,2 cm swivel casters
- Wide shelves hold a variety of equipment and supplies, up to 135 kg
- Available in various versions
Accessories information: Accessories are available for a complete system solution for housekeeping in the hospitality industry. Utility bin features a smooth easy-to-clean surface.
Informazioni per l'ordine: Attachable plastic container must be ordered separately as an accessory.
Documents
Manual Assembly instruction
Manuale
Specifications for Product Family
Specifiche
- Catalog No:
- RUBBFG409100BLA
- RUBBFG409100OWHT
- 129-1832
- 129-1833
- RUBBFG409400GRAY
- Materiale:
- PP
- PP
- PP
- PP
- PP
Specifiche
Product Family Options
Open with 3 shelves
Informazioni sul prodotto
- L×P×APesoColoreDisponibilitàPrice
- RUBBFG409100BLACapacity: 135 kg total, 45 kg per shelfCapacity: 135 kg total, 45 kg per shelf508×1032×960 mm20 kgneroLog in per disponibilità1 * 1 STSFr. 705.00Specifiche:
More
Specifications
Cat. No.RUBBFG409100BLAMaterialePP - RUBBFG409100OWHTCapacity: 135 kg total, 45 kg per shelfCapacity: 135 kg total, 45 kg per shelf508×1032×960 mm20 kgbiancoLog in per disponibilità1 * 1 STSFr. 705.00Specifiche:
More
Specifications
Cat. No.RUBBFG409100OWHTMaterialePP
Open with 4 shelves
Informazioni sul prodotto
Enclosed end panels on 3 sides
Informazioni sul prodotto
Including lockable doors and sliding drawers
Informazioni sul prodotto
- L×P×APesoColoreDisponibilitàPrice
- RUBBFG409400GRAYCapacity: 135 kg total, 45 kg per shelfCapacity: 135 kg total, 45 kg per shelf508×1032×960 mm23,6 kggrigioLog in per disponibilità1 * 1 STSFr. 1’100.00Specifiche:
More
Specifications
Cat. No.RUBBFG409400GRAYMaterialePP
Complementary Products
Informazioni sul prodotto
- Codice catalogoDescrizioneCapacitàL×P×AColoreDisponibilitàPrice
- 129-179215 L polypropylene bin; accessory for a variety of RCP utility and service carts; provides an additional compartment for storing tools and other items15,1 L432×308×267 mmneroLog in per disponibilità1 * 1 ST - SFr. 83.40
Accessories
Informazioni sul prodotto
- Codice catalogoCapacitàL×P×ADisponibilitàPrice
- 15 L polypropylene bin; accessory for a variety of RCP utility and service carts; provides an additional compartment for storing tools and other items15 L polypropylene bin; accessory for a variety of RCP utility and service carts; provides an additional compartment for storing tools and other items129-179215,1 L432×308×267 mmLog in per disponibilità1 * 1 ST - SFr. 83.40