Trolley, folding
280×260×100 mm
Codice catalogo 139-9874
Nome del fornitore: Gorr Transporttechnik GmbH
Vai a
Product Details & Documents
All purpose laboratory cart for all types of load, such as boxes, sacks, or drums. Easy to manoeuvre, even in confined spaces. Has a folding base plate to minimise storage space.
- Solid steel tube frame
- Solid rubber wheels with 125 mm
Specifiche
- L×P×A:280×260×100 mm