High leverage combination pliers
High leverage combination pliers, chrome plated, handles insulated with multi-component grips
Codice catalogo 238-5110
Nome del fornitore: KNIPEX WERKE
Vai a
Product Details & Documents
High-grade special tool steel, forged, oil-hardened. For heaviest duty. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V. With integrated forged joint axle.
- With cutting edges for soft and hard wire and piano wire
- Precision cutting edges additionally induction-hardened
- With gripping zones for flat and round material, suitable for versatile use
Length: 200 mm
Certifications: DIN ISO 5746. VDE-tested
Specifiche
- Descrizione:High leverage combination pliers, chrome plated, handles insulated with multi-component grips