Diagonal cutters
Nome del fornitore: KNIPEX WERKE
Vai a
Informazioni su questo articolo
Vanadium electric steel; forged, oil-hardened. The indispensable diagonal cutter for all-round use. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V.
- High quality material and precise workmanship for long service life
- Precision cutting edges for soft and hard wire
- Cutting edges additionally induction hardened
Certifications: DIN ISO 5749. VDE-tested.
2 varianti
Product Details & Documents
Vanadium electric steel; forged, oil-hardened. The indispensable diagonal cutter for all-round use. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V.
- High quality material and precise workmanship for long service life
- Precision cutting edges for soft and hard wire
- Cutting edges additionally induction hardened
Certifications: DIN ISO 5749. VDE-tested.
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- KNIP7005125Diagonal cutter, chrome plated, handles insulated with multi-component gripsDiagonal cutter, chrome plated, handles insulated with multi-component gripsLog in per disponibilità1 * 1 STSFr. 32.00
- 238-5310Diagonal cutter, chrome plated, handles insulated with multi-component gripsDiagonal cutter, chrome plated, handles insulated with multi-component gripsLog in per disponibilità1 * 1 STSFr. 38.00