High leverage diagonal cutter
High leverage diagonal cutter, chrome plated, handles insulated with multi-component grips
Catalog # 238-5311
Supplier: KNIPEX WERKE
Jump to:
Product Details & Documents
Chrome vanadium heavy-duty steel, forged, oil-hardened. For heaviest duty. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V. With integrated forged joint axle.
- High cutting performance with minimum effort
- Suitable for all types of wire including piano wire
- Precision cutting edges additionally induction-hardened
Length: 180 mm
Certifications: DIN ISO 5749. VDE-tested
Specifications
- Descrizione:High leverage diagonal cutter, chrome plated, handles insulated with multi-component grips