Pliers
Pliers, polished head, plastic coated handles
Codice catalogo 238-5610
Nome del fornitore: KNIPEX WERKE
Vai a
Product Details & Documents
Special tool steel, forged, oil-hardened. For heaviest duty.
- Extreme wear resistance and well balanced
- For precise workmanship
- Plastic coated handles
Length: 210 mm
Certifications: DIN ISO 9243
Specifiche
- Descrizione:Pliers, polished head, plastic coated handles