Roller for adhesive seal application
Catalog # 293-1411
Supplier: Simport Scientific
Specifications
- Descrizione:Rotolo con rullo di gomma, Amplate™
- Conf:1
Specifications
About this item
Ensure a perfect seal when using either SecureSeal™ Thermal sealing film or foil on PCR plates. Roller made of medium-hard rubber.
- Heavy-duty handle with comfortable grip reducing fatigue
- Durable construction