Scalpel handles, B3/B3L
B3, Blades No. 6/9/10/10A/11/11P/E/11/12/12D/13/14/15/15A/15C/D/15/15T/16/40 and SG3, 127 mm
Nome del fornitore: SWANN MORTON
Vai a
Product Details & Documents
Stainless steel, non sterile.
- Smooth tapered octagonal body offers precise fingertip control
- Suitable for hand surgery such as a Fasciectomy (removal of scar like tissue ), peripheral nerve repair and surgery of the tendons and joints
- B3L is longer than the standard B3 providing greater reach and depth of surgical field
Documents
Specifiche
- Per:Blades No. 6/9/10/10A/11/11P/E/11/12/12D/13/14/15/15A/15C/D/15/15T/16/40 and SG3
- Tipo:B3
- Lunghezza:127 mm
- Seq:01