Staining racks
Rack per colorazione
Catalog # 720-0096
Supplier: DIXON SCIENTIFIC
Jump to:
Product Details & Documents
Comprises two brass end-plates fitted with levelling screws and clips.
- For holding two glass rods with diameter of ∼8 to 9 mm
Ordering information: Rods are not included.
Specifications
- Descrizione:Rack per colorazione