Orders LinkContactUsLinkComponent
Rack per cuvette
Rack per cuvette
12 (2×6), 170×50×12 mm
Catalog # 634-8580
Supplier:   KARTELL
Rack per cuvette
Catalog # 634-8580
Supplier:  KARTELL
Rack per cuvette
Rack per cuvette
12 (2×6), 170×50×12 mm
Price
SFr. 6.00
Log in for availability
1 * 1 ST
Product Details & Documents
PE.
Rack per dodici cuvette 10×10 mm.
Documents
Specifications
  • Per:
    12 (2×6)
  • L×P×A:
    170×50×12 mm