Pipette Tips, EasyLoad® system
EasyLoad® is a simple, fast and space-saving reload system for micropipette tips.
- Graduated tips, sterile or non sterile
- EasyLoad® racks are autoclavable
- Recyclable tips and racks
: EasyLoad® 200 system also available as a starter kit.
Frequently Bought Together