Footrest, Swing
Poggiapiedi per il massimo relax, grigio chiaro, 490×350×190 mm, 2,4 kg
Codice catalogo 139-9923
Nome del fornitore: WEDO WERNER DORSCH
Vai a
Product Details & Documents
The footrest is produced from strong polystyrol plastic and features a covered compartment for foot control of dictating machines, copyholders, etc.
- Adjustable in height and inclination
- Recess for foot switch
- The ribbed platform ensures a firm grip and avoids slipping
Certifications: In compliance with DIN standard 4556, GS tested.
Specifiche
- Peso:2,4 kg
- Colore:grigio chiaro
- Descrizione:Poggiapiedi per il massimo relax
- L×P×A:490×350×190 mm