Guanti isolanti, ActivArmr® 78-101
Supplier: Ansell
Certificates
About this item
These ambidextrous, light and stretchable thermal protection gloves are made of special hollow-core Thermolite® fibres which provide a very good thermal insulation barrier between the skin and the surrounding environment.
- Dry faster than cotton: Fabric efficiently eliminates moisture from the skin, thus ensuring the wearer's warmth and comfort
- Automatic knit technology construction permits longer, more comfortable wear
- No seams to rub or chafe: Designed to avoid any discomfort or skin irritation
- Washable at maximum 40 °C
The gloves can be used either as primary gloves or undergloves to protect against cold wherever required. Ideal for outdoor workers.
Certifications: EN 388:2016 (214XA), EN 407 (X1XXXX), EN 511 (100), cat. III
Specifications
- Catalog No:
- 112-0213
- 112-0212
- Ambidestro/mano specifico:
- ambidestro
- ambidestro
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- Calibro:
- 13
- 13
- Tipo polsino:
- Maglia elastica
- Maglia elastica
- Norma EN:
- EN 388:2016 (214XA), EN 407 (X1XXXX), EN 511 (100)
- EN 388:2016 (214XA), EN 407 (X1XXXX), EN 511 (100)