Guanti in lattice, sterili, per camera bianca, BioClean Alpha™
Supplier: Nitritex
Certificates
About this item
Guanti in lattice con bordino e forma anatomica.
- Sterile, powder-free, textured surface
- Extra thick cuff to reduce the incidence of tearing when donning
- Protein level less than 50 µg/g
- Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
- Cuff marking: Ink Print Size and L or R
- AQL 1,5
- Standard compliance: PPE, Cat III EN ISO 374 Type B
Per camere bianche di classe 10, ISO 4 o EU GMP grado A.
Length: 300 mm
Thickness: 0,18 mm (cuff); 0,21 mm (palm); 0,23 mm (fingers)
Colour: White
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009 and EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009 and EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Packaging: un paio con polsini in involucro a portafoglio interno; un involucro a portafoglio interno per bustina a strappo EasyTear™ in PE; 10 paia per sacchetto in PE esterno sigillato; 20 sacchetti esterni per confezione (200 paia).
Specifications
- Catalog No:
- 112-3059
- 112-3058
- 112-3057
- 112-3056
- 112-3055
- 112-3061
- 112-3060
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- monouso
- monouso
- monouso
- monouso
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Classe di cleanroom:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Tipo polsino:
- Arrotolato
- Cuff_Style-21391103|Cuff_Style-21391099
- Arrotolato
- Arrotolato
- Arrotolato
- Arrotolato
- Arrotolato
- Spessore polsino:
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- Spessore delle dita:
- 0,24 mm
- 0,24 mm
- 0,24 mm
- 0,24 mm
- 0,24 mm
- 0,24 mm
- 0,24 mm
- Spessore palmo:
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- Resistenza tessile:
- Min. 14 Mpa
- Min. 14 Mpa
- Min. 14 Mpa
- Min. 14 Mpa
- Min. 14 Mpa
- Min. 14 Mpa
- Min. 14 Mpa
- Norma EN:
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374