About this item
Soluzione pratica e conveniente per controllo di sterilità negli isolatori.
- Economical
- Ideal for aqueous solutions
- Individually bagged
MicroFunnel™ ST filter funnels are ideal for testing any aqueous solution for microbial contamination using the principles of Membrane Filter (MF) technique.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications or 0,2 and 0,45 µm Supor® (polyethersulfone) membrane to minimise binding of inhibitory substances. Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
Individually bagged funnels are packaged within an overpack bag. The double bagged design makes them easy to transfer into isolators or more convenient to wipe down for use in cleanrooms and hoods. Chose from a variety of membranes and pore sizes to best suit your testing needs.
300 ml: 87×89 mm with cover
Specifications
- Catalog No:
- PALL4851
- 516-1237
- 516-9096
- PALL4857
- 515-0181
- 516-9095
- 516-1236
- 516-9094
- 516-9097
- Capacità:
- 100 ml
- 300 ml
- 100 ml
- 300 ml
- 300 ml
- 300 ml
- 100 ml
- 100 ml
- 300 ml
- Superficie filtrante effettiva:
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- Dimensioni:
- 76H×61W mm
- 89H×87W mm
- 76H×61W mm
- 89H×87W mm
- 89H×87W mm
- 89H×87W mm
- 76H×61W mm
- 76H×61W mm
- 89H×87W mm
Specifications
Related Information
Certificates
Search by Lot NumberMembrane Filter Funnels, MicroFunnel™, PALL Laboratory
Application Notes
Pall Laboratory Microbiology QC Products
Product Literature
PALL4811ME_TDS_EN