About this item
Semplificazione dei processi di concentrazione, desalinizzazione e sostituzione del tampone su scala di laboratorio (fino a 1 litro).
- High product recovery
- Minimal sample loss
- Greater efficiency
- Gentle processing
- Plug and play processing
- Scalable operation
Versatile, self-contained capsules include all fittings and tubing to simplify installation into filtration set-ups and will work with any laboratory pump. Dual-pressure gauges allow trans-membrane pressure (TMP) differential calculations.
Capsules feature glass-reinforced polypropylene housing, polypropylene screens, housing sealing with fittings, polyethylene membrane plate/filtrate channels, and an internal gasket made of ethylene propylene elastomer.
The Minimate™ TFF system, designed to work with the Minimate™ TFF capsule, is ideal for a variety of tangential flow filtration applications in biotechnology, molecular biology, protein chemistry, microbiology and immunology. This ready to use system can concentrate and desalt proteins, peptides or nucleic acid sample volumes of up to 1 L or more efficiently concentrate samples to as little as 5 ml. System also features gentle and efficient processing, easy cleaning and maintenance, compact design, and high concentration factors.
The optimised flow path design of the capsules coupled with low protein binding Omega™ (modified polyether sulfone) ultrafiltration membrane minimise non specific binding. Capsules allow concentration and desalting processes to be performed on the same system with minimal user intervention. Several Minimate™ TFF capsules may be connected in parallel for increased membrane area.
- Concentrate viruses or gene therapy vectors
Specifications
- Catalog No:
- 516-1085
- Superficie filtrante effettiva:
- 50 cm2
- Dimensioni:
- 307×482×208 mm
- Pressure:
- 4.1 bar (410 kPa, 60 psi)