Universal container, PP, with PE screw cap
Catalog # 216-2053
Supplier: Brand
Specifications
- Ø est:65 mm
- Versione:sterile (yellow cap)
- Capacità:100 ml
- Altezza:75 mm
- Seq:02
- Conf:240
Specifications
About this item
PP, translucent with green PE screw cap.
- Non sterile or sterile
- Graduated to 100 ml
For hygienic fluid sample handling.