Anion exchange HPLC columns, RCX-30
Supplier: HAMILTON BONADUZ
Certificates
About this item
RCX-30 carbohydrate analysis columns are designed for the isocratic or gradient separation of carbohydrates. When an isocratic method is used with a conductivity, refractive index, ultraviolet, or pulsed amperometric detector (PAD), mono and disaccharides such as glucose, fructose, sucrose and lactose can be quickly determined.
- Greater exchange capacity than that of the PRP-X100
- Samples with two or three carbohydrates can be quickly separated isocratically
- More complex samples require gradient elution
Specifications
- Catalog No:
- 554-0212
- 554-1248
- 554-1247
- 552-0350
- 554-0470
- Pressione max.:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- Range di pH:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- Limite di temperatura:
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)