HPLC columns, Develosil® ODS-UG
Supplier: HICHROM
Certificates
About this item
Develosil® ODS-UG is the most stable column under alkaline conditions (pH 1 to 10).
- Spherical porous silica
Specifications
- Catalog No:
- 554-7681
- Carico di carbonio:
- 18%
- Con endcapping:
- Sì
- Area superficiale:
- 300 m²/g