MPLC cartridge columns, Brownlee™
PERC07110016
- Formato della colonna:Analitica
- Ø int.:4,60 mm
- Dimensioni particelle:5 µm
- Dimensioni pori:80 Å
- Descrizione:Spheri-5®
- Lunghezza:100 mm
- Con endcapping:Sì
- Area superficiale:180 m²/g
- Carico di carbonio:11%
- Fase:RP-18
- Conf:1
The MPLC cartridge system allows direct coupling of an analytical and NewGuard™ cartridge without introduction of any dead volume. A specific cartridge holder (30, 100, 220 mm) is required for each cartridge length.
- Easy changing with fingertight cartridge
- Avoids disconnecting of tubing from the LC system