Orders LinkContactUsLinkComponent
Raschig rings, Quickfit®
Raschig ring, 9 mm, 9 mm
  BIBBFC8/09
 :   SciLabware
Raschig rings, Quickfit®
  BIBBFC8/09
Raschig rings, Quickfit®
Raschig rings, Quickfit®
Raschig ring, 9 mm, 9 mm
SFr. 417.00
1 * 1 ST
PYREX®, borosilicate glass.
  • For use with Quickfit® plain fractionating columns
  • Good chemical resistance
  • Good thermal resistance
Short lengths of glass tubing. Manufactured from borosilicate glass for contamination-free performance.
 : Supplied in 1 litre bags.
  • Ø est:
    9 mm
  • Descrizione:
    Raschig ring
  • Altezza:
    9 mm
  • Seq:
    02