Raschig rings, Quickfit®
Raschig ring, 9 mm, 9 mm
BIBBFC8/09
PYREX®, borosilicate glass.
- For use with Quickfit® plain fractionating columns
- Good chemical resistance
- Good thermal resistance
Short lengths of glass tubing. Manufactured from borosilicate glass for contamination-free performance.
: Supplied in 1 litre bags.
- Ø est:9 mm
- Descrizione:Raschig ring
- Altezza:9 mm
- Seq:02