Tute, Secutex® pro 5/6
Nome del fornitore: Zellstoff-Vertriebs-GmbH & Co. KG
Informazioni su questo articolo
This particulate resistant disposable overall made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals. The overall comes complete with a protective flap over the zip, elasticated hood, back, sleeves and ankles.
- Generous cut provides excellent freedom of movement, making it extremely comfortable to wear
- Permeable to air and water vapour
- Flexible, lint-free, abrasion resistant and anti-static material
Colour: White.
Type 5/6, EN 1149, Cat. III/CE 01
: CE 0624, Codice III, Tipo 5 + 6.
4 varianti
Product Details & Documents
This particulate resistant disposable overall made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals. The overall comes complete with a protective flap over the zip, elasticated hood, back, sleeves and ankles.
- Generous cut provides excellent freedom of movement, making it extremely comfortable to wear
- Permeable to air and water vapour
- Flexible, lint-free, abrasion resistant and anti-static material
Colour: White.
Type 5/6, EN 1149, Cat. III/CE 01
: CE 0624, Codice III, Tipo 5 + 6.
Specifications for Product Family
Specifiche
- Catalog No:
- 113-0657
- 113-0650
- 113-0651
- ZVGA30445-90
- Antistatico:
- +
- +
- +
- +
- Applicazioni per abiti:
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.113-0657Antistatico+Applicazioni per abitiChemical protection|Laboratory|Industrial - Specifiche:
More
Specifications
Cat. No.113-0650Antistatico+Applicazioni per abitiChemical protection|Laboratory|Industrial - Specifiche:
More
Specifications
Cat. No.113-0651Antistatico+Applicazioni per abitiChemical protection|Laboratory|Industrial - Specifiche:
More
Specifications
Cat. No.ZVGA30445-90Antistatico+Applicazioni per abitiChemical protection|Laboratory|Industrial