Overalls, Proshield® 4 Practik, model CHO5
N° de catalogue 113-3889
Fournisseur: DuPont
Spécifications
- Taglia dell'abbigliamento:M
- Applicazioni per abiti:General Purpose,Industrial
- Colore:bianco
- Seq:01
- Modello:ProShield® 4 Practik
- Conf:25
- Peso:41,5 g/m²
Spécifications
À propos de cet article
These garments are made of a proprietary DuPont micro-perforated non woven polyethylene fabric. ProShield® 4 Practik overalls are comfortable because the material is micro-perforated to allow the unrestricted passage of air and water vapour.
- Hood, nylon zipper and flap
- Elasticated wrists, ankles, face and waist (back)
- Stitched seams
- Antistatically treated for comfort (not acc. EN 1149-5)
- Not CE certified as chemical protective clothing
Ideal workwear for do-it-yourself, general maintenance and cleaning, manufacturing and other non hazardous applications as well as a re-usable garment for visitors.
Colour: white
Weight: 41,5 g/m²
Certifications: Cat. I