Orders LinkContactUsLinkComponent
Clamps, 4-prong
 :   BOCHEM
Clamps, 4-prong
 :  BOCHEM
Clamps, 4-prong
Clamps, 4-prong
Acciaio inox.
  • Without bosshead for fixing to rod
  • Span width independently adjustable by two regulating screws
Product Family Options
  • Larghezza di serraggio
  • 30 - 100 mm
    1 * 1 ST
    1 * 1 ST - 131.00 SFr.
    131.00 SFr.
    1 * 1 ST
       131.00 SFr.
  • 50 - 150 mm
    1 * 1 ST
    1 * 1 ST - 144.00 SFr.
    144.00 SFr.
    1 * 1 ST
       144.00 SFr.