Orders LinkContactUsLinkComponent
Inoculating loops with handle | Avantor
Inoculating loops with handle
Katalognr. 631-7144
Anbieter:   HAMMACHER KARL
Inoculating loops with handle
Katalognr. 631-7144
Anbieter:  HAMMACHER KARL
Inoculating loops with handle
Inoculating loops with handle
Preis
52.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Spatula-shaped end.
Loop Ø: 4 mm
Length: 210 mm