Accessories for inoculating loops and needles
631-2240
- Descrizione:Stand for inoculating loops and needles
- Conf:1
PP. For storage of 6 needles and inoculation loops. Ø 120 mm.
- Round
- With 2×6 holes for Ø 7 and 9 mm
- With central hole for loop holders