Orders LinkContactUsLinkComponent
SampliQ SPE cartridges
SampliQ SPE cartridges
SampliQ SPE cartridges
Certificates
SampliQ SPE cartridges
14 Options Available Below
Product Details & Documents
High quality SampliQ SPE (Solid Phase Extraction) products help the user confidently extract and concentrate samples from complex matrices, ensuring fast, accurate, and reproducible results from the very first step. The SampliQ line includes polymers, silica and non-silica options to meet user's specific SPE needs.
  • Improve and simplify separations
  • Increase column lifetime
  • Improve detection limits
SampliQ SPE products are manufactured in the US to ISO-9001 standards.
Packaging: To ensure product integrity all SampliQ products are packaged with triple lamination and include a certificate of performance.
Product Family Options
With silica
Product Information
  • Descrizione
    Formato della colonna
    Fase
    Dimensioni pori
    Dimensioni particelle
    Capacità
    Peso letto
    Availability
    Price
  • Cartuccia
    C18
    60 Å
    45 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • Cartuccia
    C18
    60 Å
    45 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • Cartuccia
    C8
    60 Å
    45 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • Cartuccia
    C8
    60 Å
    45 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • Cartuccia
    NH₂
    60 Å
    45 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • Cartuccia
    SAX
    60 Å
    45 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • Cartuccia
    SAX
    60 Å
    45 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • SPE ion exchange, anion Si-SCX
    Cartuccia
    SCX
    60 Å
    45 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • Cartuccia
    SCX
    60 Å
    45 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • Cartuccia
    SCX
    60 Å
    45 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • Cartuccia
    Si
    60 Å
    45 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • Cartuccia
    Si
    60 Å
    45 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
With other sorbents
Product Information
  • Descrizione
    Formato della colonna
    Fase
    Dimensioni pori
    Dimensioni particelle
    Capacità
    Peso letto
    Availability
    Price
  • Cartuccia
    Florisil PR
    200 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • Cartuccia
    Florisil PR
    200 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
This product has been blocked by your organization. Please contact your purchasing department for more information