Contenitore di sicurezza per il trasporto, Nalgene®
Nome del fornitore: Thermo Fisher Scientific
Certificati
Informazioni su questo articolo
For storing and transporting 2,5 or 4 litre chemical or acid bottles. Also suitable for bottles with handles on the neck.
LDPE, with PC lid.
- Protective cover is closed firmly around the bottle by the carrying handle when it is being carried
- Transparent plastic cap protects the bottle closure, which is visible through the cap, allowing identification of colour coded closures on acid bottles
- Cap can be unscrewed, the bottle does not have to be taken out of the container to be able to use its contents
Specifiche
- Catalog No:
- 216-4426
- 216-4427
- Environmentally Preferable:
- Produzione a basso impatto
- Produzione a basso impatto
- Certificazione ambientale:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)