Bottiglie per burette automatiche
Nome del fornitore: Brand
Vai a
Informazioni su questo articolo
Borosilicate glass 3.3.
- Very good temperature resistance
- Very good chemical resistance
- Amber coloured for light sensitive substances
- Socket size NS 29732
Reservoir bottle for automatic burette available in clear glass or amber glass.
2 varianti
Product Details & Documents
Borosilicate glass 3.3.
- Very good temperature resistance
- Very good chemical resistance
- Amber coloured for light sensitive substances
- Socket size NS 29732
Reservoir bottle for automatic burette available in clear glass or amber glass.