Buckets, square, Azlon®
500 ml, bianco, 14 L
Codice catalogo 216-2811
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
HDPE, white or black.
- Square shape allows economical use of space
- Handle grip for easy lifting and carrying
- Lids available with moulded inset handle
Specifiche
- Capacità:14 L
- Colore:bianco
- Divisione:500 ml