Dispenser for Small Items
Dispenser per minuteria, 438×235×305 mm
Codice catalogo 113-0033
Nome del fornitore: AK
Vai a
Product Details & Documents
Clear acrylic.
For storing and dispensing disposable items such as overshoes, gloves or hoods and many other laboratory utensils.
- Four compartments with hinged lid at filling openings and removal points
- Easy to clean
- Designed for use as wall or bench model
Specifiche
- Descrizione:Dispenser per minuteria
- L×P×A:438×235×305 mm