Compressed air sprayer
1000 ml, con beccuccio corto
Codice catalogo 215-6575
Nome del fornitore: KLAGER PLASTIK
Vai a
Product Details & Documents
HDPE.
- Good chemical resistance, good temperature resistance
- With screw threaded bottle
- Choose between fine mist or spray by adjusting the spray nozzle
For aqueous solutions. Air is pumped into the container using a pump lever (depending on filling level approx. 20 to 30 pump actions). The spray mist or jet can be easily regulated with the spray nozzle.
Specifiche
- Tipo:con beccuccio corto
- Capacità:1000 ml