ContactUsLinkComponent
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up
Nome del fornitore:  Thermo Fisher Scientific
Certificati
undefined
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up
Nome del fornitore:  Thermo Fisher Scientific
Specifiche
  • Catalog No:
  • BARNCE8120BB
  • BARNCE8140BB
  • BARNCE8180BB
  • BARNCE8194BB
  • Modello:
  • CryoExtra 20 BB
  • CryoExtra 40 BB
  • CryoExtra 80 BB
  • CryoExtra 94 BB
  • Capacità:
  • 463 L
  • 797 L
  • 1745 L
  • 1770 L
  • L×P×A est.:
  • 813×813×1651 mm
  • 1245×1295×1702 mm
  • 1524×1542×1732 mm
  • 1524×1524×1859 mm
  • Peso, pieno di LN₂:
  • 348 kg
  • 327 kg
  • 781 kg
  • 781 kg