About this item
The SL2001 set by ebro, including the EBI 12-T220, EBI 12-T441, and EBI 12-TP231 loggers, is tailored for validation of washer-disinfectors (RDG) in healthcare and industrial settings, ensuring compliance with cleaning and disinfection standards.
- Complete validation set for washer-disinfectors
- Includes EBI 12-T220, EBI 12-T441, and EBI 12-TP231 loggers
- Monitors temperature and pressure with high precision
- Ensures hygiene compliance in healthcare and industrial sectors
- Robust, IP-rated devices for durability in harsh environments
- Factory calibration certificate
The SL2001 set from ebro is designed specifically for comprehensive validation of washer-disinfectors (RDG) in healthcare, laboratory, and industrial applications, ensuring rigorous compliance with cleaning and disinfection standards. The set includes three high-precision loggers: the EBI 12-T220 and EBI 12-T441 for accurate temperature tracking, and the EBI 12-TP231 for combined temperature and pressure monitoring. Each device offers robust data collection with high accuracy, enabling detailed cycle analysis and quality control. The loggers are IP-rated for durability, making them reliable tools for routine validations in demanding environments.
Logger dimensions: Ø 48×24 mm H
Specifications
- Catalog No:
- EBRO1340-6068
- EBRO1250-2001
- Modello:
- —
- —
- Sensore:
- —
- —
- Allarme:
- —
- —
- Classe di protezione:
- —
- —
- Intervalli di registrazione:
- —
- —
- Peso:
- —
- Memoria dati - N. letture:
- —
- —
- Batteria:
- —
- —
Specifications
Related Information
Data logger Monitoring of Cold Chain and Production Data logger Validation and Process Monitoring
Application Notes
Professional Data Loggers