Masterflex® Miniflex® OEM Panel Mount Pumps Systems
Informazioni su questo articolo
Designed for easy integration into OEM designs wherever precision fluid handling is needed.
- Precise flowrates up from 0,12 to 390 ml/min (3 to 300 min⁻¹)
- Rugged sturdy design with durable pump head materials
- Pump head accepts continuous lengths of tubing for clean flow paths
- Automatic tubing retention makes tube loading quick and easy
- IP65 Ingress protection; removable head for spray and wipe down cleanability
- High efficiency and dependable brushless DC motor
- I/O control features include analogue speed control, direction, tach output, open head sensor alarm, and general alarm
- Reduced pulsation options available with 4 or 6 roller pump heads
- Multichannel capability with dual channel pump head option
- More than 20 tubing formulations to support chemical and compliance requirements
- Customisable options available
Applications: Biopharma production, testing and research; Laboratory instrumentation; Air and liquid sampling devices; Food and beverage dispensing; Ink and printing; Industrial fluid transfer; Wastewater management.
Miniflex® panel mount systems include everything needed to incorporate Masterflex® peristaltic pumping into OEM product designs. These models feature an IP65 ingress protection to ensure proper seal during cleaning and for use in critical environments. The BLDC motors provide a combination of efficiency and long term durability. Miniflex® pump head allows for fast tubing changes with up to four tubing sizes.
: UL 61010-1, CSA, CE (EN 61010-1 Low Voltage, EN 61326-1 EMC Directive, EN ICE 63000 ROSH Directive, EN 809 Machinery Directive), ISO 9001.
: For use with Masterflex® Pump tubing, sold separately. Two channel pump head options are available and pump system is rated for use with all Masterflex® tubing formulations. Choose four roller pump head design for reduced pulsation. Customised options are available.
3 varianti
Product Details & Documents
Designed for easy integration into OEM designs wherever precision fluid handling is needed.
- Precise flowrates up from 0,12 to 390 ml/min (3 to 300 min⁻¹)
- Rugged sturdy design with durable pump head materials
- Pump head accepts continuous lengths of tubing for clean flow paths
- Automatic tubing retention makes tube loading quick and easy
- IP65 Ingress protection; removable head for spray and wipe down cleanability
- High efficiency and dependable brushless DC motor
- I/O control features include analogue speed control, direction, tach output, open head sensor alarm, and general alarm
- Reduced pulsation options available with 4 or 6 roller pump heads
- Multichannel capability with dual channel pump head option
- More than 20 tubing formulations to support chemical and compliance requirements
- Customisable options available
Applications: Biopharma production, testing and research; Laboratory instrumentation; Air and liquid sampling devices; Food and beverage dispensing; Ink and printing; Industrial fluid transfer; Wastewater management.
Miniflex® panel mount systems include everything needed to incorporate Masterflex® peristaltic pumping into OEM product designs. These models feature an IP65 ingress protection to ensure proper seal during cleaning and for use in critical environments. The BLDC motors provide a combination of efficiency and long term durability. Miniflex® pump head allows for fast tubing changes with up to four tubing sizes.
: UL 61010-1, CSA, CE (EN 61010-1 Low Voltage, EN 61326-1 EMC Directive, EN ICE 63000 ROSH Directive, EN 809 Machinery Directive), ISO 9001.
: For use with Masterflex® Pump tubing, sold separately. Two channel pump head options are available and pump system is rated for use with all Masterflex® tubing formulations. Choose four roller pump head design for reduced pulsation. Customised options are available.
Specifications for Product Family
Specifiche
- Catalog No:
- MFLX77222-10
- MFLX77222-12
- MFLX77222-14
- Modello:
- 77222-10
- 77222-12
- 77222-14
- Accuratezza:
- ±0,1%
- ±0,1%
- ±0,1%
- Tipo di controllo:
- Remote 4 - 20 mA
- Remote 0 - 10 V input
- Remote 4 - 20 mA
- Pressione max.:
- 1 bar (15 psi) @ max flow
- 1 bar (15 psi) @ max flow
- 1 bar (15 psi) @ max flow
- N. di canali:
- 1
- 1
- 1
- Pump series:
- L/S
- L/S
- L/S
Specifiche
Product Family Options
Pump systems
Informazioni sul prodotto
- Massima velocità del flussoPotenzaAltezzaLunghezzaDisponibilitàPrice
- MFLX77222-10Miniflex® OEM panel mount pumps sytem with 3-roller pump head, 4 - 20 mA input, Tubing sizes accepted:Miniflex® OEM panel mount pumps sytem with 3-roller pump head, 4 - 20 mA input, Tubing sizes accepted:390 ml/min24 VDC130 mm223 mmLog in per disponibilità1 * 1 STSFr. 1’630.00Specifiche:CE CompliantUL recognized
More
Specifications
Cat. No.MFLX77222-10Modello77222-10Accuratezza±0,1%Tipo di controlloRemote 4 - 20 mAPressione max.1 bar (15 psi) @ max flowN. di canali1Pump seriesL/SCE CompliantUL recognized - MFLX77222-12Miniflex® OEM panel mount pumps sytem with 3-roller pump head, 0 - 10 mA input, Tubing sizes accepted:Miniflex® OEM panel mount pumps sytem with 3-roller pump head, 0 - 10 mA input, Tubing sizes accepted:390 ml/min24 VDC130 mm223 mmLog in per disponibilità1 * 1 STSFr. 1’630.00Specifiche:CE CompliantUL recognized
More
Specifications
Cat. No.MFLX77222-12Modello77222-12Accuratezza±0,1%Tipo di controlloRemote 0 - 10 V inputPressione max.1 bar (15 psi) @ max flowN. di canali1Pump seriesL/SCE CompliantUL recognized - MFLX77222-14Miniflex® OEM panel mount pumps sytem with 4-roller pump head, 4 - 20 mA input, Tubing sizes accepted:Miniflex® OEM panel mount pumps sytem with 4-roller pump head, 4 - 20 mA input, Tubing sizes accepted:315 ml/min24 VDC130 mm223 mmLog in per disponibilità1 * 1 STSFr. 1’630.00Specifiche:CE CompliantUL recognized
More
Specifications
Cat. No.MFLX77222-14Modello77222-14Accuratezza±0,1%Tipo di controlloRemote 4 - 20 mAPressione max.1 bar (15 psi) @ max flowN. di canali1Pump seriesL/SCE CompliantUL recognized
Accessories
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, PharMed® BPT, L/S 13; 25 ftL/S® Precision Pump Tubing, PharMed® BPT, L/S 13; 25 ftMFLX06508-130,80 mm (0,031")PharMed® BPTLog in per disponibilità1 * 25 Piedi - 142.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 14; 25 ftL/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 14; 25 ftMFLX96410-141,60 mm (0,063")Platinum-cured siliconeLog in per disponibilità1 * 25 Piedi - 118.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 13; 25 ftL/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 13; 25 ftMFLX96410-130,80 mm (0,031")Platinum-cured siliconeLog in per disponibilità1 * 25 Piedi - 95.00 SFr.
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement 25 pin I/O cable for panel mount pump systems (1 m)Replacement 25 pin I/O cable for panel mount pump systems (1 m)MFLX77222-92Log in per disponibilità1 * 1 ST - 107.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, PharMed® BPT, L/S 25; 25 ftL/S® Precision Pump Tubing, PharMed® BPT, L/S 25; 25 ftMFLX06508-254,80 mm (0,189")PharMed® BPTLog in per disponibilità1 * 25 Piedi - 209.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 16; 25 ftL/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 16; 25 ftMFLX96410-163,10 mm (0,122")Platinum-cured siliconeLog in per disponibilità1 * 25 Piedi - 114.00 SFr.
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement 24 DC power cable with ground (1 m)Replacement 24 DC power cable with ground (1 m)MFLX77222-90Log in per disponibilità1 * 1 ST - 107.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 25; 25 ftL/S® Precision Pump Tubing, Platinum-Cured Silicone, L/S 25; 25 ftMFLX96410-254,80 mm (0,189")Platinum-cured siliconeLog in per disponibilità1 * 25 Piedi - 150.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, PharMed® BPT, L/S 14; 25 ftL/S® Precision Pump Tubing, PharMed® BPT, L/S 14; 25 ftMFLX06508-141,60 mm (0,063")PharMed® BPTLog in per disponibilità1 * 25 Piedi - 183.00 SFr.
Informazioni sul prodotto
- Codice catalogoØ int.tuboMateriale del tuboDisponibilitàPrice
- L/S® Precision Pump Tubing, PharMed® BPT, L/S 16; 25 ftL/S® Precision Pump Tubing, PharMed® BPT, L/S 16; 25 ftMFLX06508-163,10 mm (0,122")PharMed® BPTLog in per disponibilità1 * 25 Piedi - 183.00 SFr.