Orders LinkContactUsLinkComponent
Fluid Sampling Kit
Fluid Sampling Kit
Fluid sampling kit, non sterile, contains PTFE hose(s), polystyrene monitor(s), L×W×H: 368×235×125 mm, Acciaio inossidabile
Catalog # MILFXX6403730
Supplier:   Merck
Fluid Sampling Kit
Catalog # MILFXX6403730
Supplier:  Merck
Fluid Sampling Kit
Fluid Sampling Kit
Fluid sampling kit, non sterile, contains PTFE hose(s), polystyrene monitor(s), L×W×H: 368×235×125 mm, Acciaio inossidabile
Price
SFr. 3’820.00
Log in for availability
1 * 1 ST
Product Details & Documents
Fluid sampling kit for sampling turbine fuels, water or other liquids through Millipore contamination analysis monitors.
  • Sturdy carrying case and stainless steel sampling assembly provide years of in-field service
  • Fluid Sampling Kit includes 12 Tenite contamination monitors
The Fluid Sampling Kit is a complete solution for field-based fluid collection and contamination analysis. The stainless steel sampler assembly plugs into a quick-release valve installed in the system line, which allows a measured quantity of liquid to pass through the monitor. A valved syringe serves as a pump to remove residual liquid from the monitor. Kit is compatible with our 37 mm contamination monitors and matched-weight monitors.
Specifications
  • Descrizione:
    Fluid sampling kit, non sterile, contains PTFE hose(s), polystyrene monitor(s), L×W×H: 368×235×125 mm
  • Materiale:
    Acciaio inossidabile
Accessories
Product Information
  • Catalog #
    Descrizione
    Conf
    Availability
    Price
  • MILFASTM03701
    Colour standards
    1
    Log in for availability
    1 * 1 ST - SFr. 239.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MILFXX3002502
    Filter holder gasket, PTFE
    Log in for availability
    1 * 10 ST - SFr. 101.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MILFXX3002510
    Filter holder support screen, stainless steel
    Log in for availability
    1 * 1 ST - SFr. 44.92