EZ-Fit™ Manifold
Catalog # MILFEZFITMIC03
Supplier: Merck
Specifications
- Descrizione:EZ-Fit™ Manifold, 3-place for Microfil® funnels and membranes
- Materiale:Acciaio inossidabile
- Seq:01
- Conf:1
Specifications
About this item
EZ-Fit™ Manifold for universal laboratory filtration.
- Unique design for easy cleaning and decontamination
- Different filtration heads for both reusable and disposable filtration devices
- Quick-fit connectors for the vacuum tubing
- Low height for ease of use in laminar flow hoods
- Each component can be removed quickly and easily without any additional tools
- A check valve is integrated into the tubing connection to prevent liquid back-flow from the vacuum system
- The 2-way valve prevents the user from accidentally touching the filtration head while opening or closing it
The new EZ-Fit™ Manifold design eliminates sources of contamination that can cause false positive results: all internal areas are accessible and can be cleaned efficiently and all components can be autoclaved.