Auftragserfassung
Switzerland
ContactUsLinkComponent
SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
Catalog # MILFTISNAP2MM
Supplier:  Merck
undefined
SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
Catalog # MILFTISNAP2MM
Supplier:  Merck
Specifications
  • Descrizione:
    SNAP i.d.2 Protein detection system, Mini and Midi
  • Seq:
    02
  • Conf:
    1
Specifications
About this item
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Ordering information: This system includes the SNAP i.d. 2.0™ base, a mini blot holding frame,a midi blot holding frame, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.