HPLC Columns, Reprospher 100 C18
Supplier: DR MAISCH
New Product
Certificates
About this item
Reprospher media are based on a 100 Å ultrapure silica and are fully scalable from UPLC to preparative and process scale applications.
Surface area: 350 m²/g
Phase/modification: C18 + carboxylic acids
pH range: 2 to 8
Specifications
- Catalog No:
- MAISRS15.ARG.S2520
- MAISRS15.9AC.S2520
- Phase name:
- C18, ODS
- C18, ODS
- Carico di carbonio:
- 16 %
- 16 %
- Range di pH:
- 2 - 8
- 2 - 8
- Area superficiale:
- 350 m²/g
- 350 m²/g
- Codice USP:
- L1
- L1