Pehazell® Clean Cellulose Wadding
Nome del fornitore: HARTMANN PAUL
Vai a
Informazioni su questo articolo
The cellulose wadding with high absorbency. Highly bleached or unbleached cellulose wadding with high absorbency capacity.
- High absorbency
- Suitable as absorptive and padding material
Unbleached cellulose is only used for padding and cleaning purposes in general care.
Available as rolls or layers.
4 varianti
Product Details & Documents
The cellulose wadding with high absorbency. Highly bleached or unbleached cellulose wadding with high absorbency capacity.
- High absorbency
- Suitable as absorptive and padding material
Unbleached cellulose is only used for padding and cleaning purposes in general care.
Available as rolls or layers.
Product Family Options
Informazioni sul prodotto
- ConfezioneDisponibilitàPrice
- HART9130610Cleaning rolls highly bleached, 360 mmCleaning rolls highly bleached, 360 mm1000 g/pack, 6 packs/boxLog in per disponibilità1 * 1 kg40.50 SFr.
- HART9130730Cleaning layers unbleached, 185×190 mmCleaning layers unbleached, 185×190 mm15 kg/pack, 1 pack/boxLog in per disponibilità1 * 15 kg159.00 SFr.
- HART9130640Cleaning layers highly bleached, 370×570 mmCleaning layers highly bleached, 370×570 mm3×5 kg/pack, 1 pack/box, 12 pack/palletLog in per disponibilità1 * 15 kg278.00 SFr.
- HART9130760Cleaning layers unbleached, 370×570 mmCleaning layers unbleached, 370×570 mm15 kg/pack, 1 pack/box, 16 packs/palletLog in per disponibilità1 * 15 kg192.00 SFr.