
Zinco 0,05 mol/L
Catalog # ACSDCH8006033.1L
Supplier: CPACHEM
New Product
CAS Number:
Specifications
- Conf:1 L
- Seq:5037
Specifications
About this item
:
:65.39 g/mol
Boiling Pt:100 °C
Density:1 g/cm³
:GHS07
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291