Beaded Edge, PTFE Faucet
NEUB1195-55-25250
New Product
- Descrizione:Beaded edge, PTFE faucet
- Conf:1
Beaded edge PTFE faucet with punctures.
- Borosilicate 3.3
- PTFE tap NS 14,5/2,5 mm
Internal diameter: Ø 25 mm
Length: 250 mm
Tube: Ø 28×1,4 mm
Beaded edge PTFE faucet with punctures.
Internal diameter: Ø 25 mm
Length: 250 mm
Tube: Ø 28×1,4 mm