Intelsius® PharmaTherm Temperature Controlled Packaging
Utilises pharmachill cool packs are hard wearing and durable re-usable coolant, reducing costs and landfill waste.
2 to 8 °C systems qualified against the Intelsius high performance quad profile with stress peaks and overlapping mid season temperatures, the PharmaTherm offers robust performance for all season shipping and peace of mind.
15 to 25 °C systems qualified against Intelsius controlled room temperatures profiles for ambient shipments. GDP compliant systems.
Utilises pharmachill cool packs are hard wearing and durable re-usable coolant, reducing costs and landfill waste.
2 to 8 °C systems qualified against the Intelsius high performance quad profile with stress peaks and overlapping mid season temperatures, the PharmaTherm offers robust performance for all season shipping and peace of mind.
15 to 25 °C systems qualified against Intelsius controlled room temperatures profiles for ambient shipments. GDP compliant systems.
Product Family Options
High perfomance quad profiles
- DescrizioneCapacitàEsterno L×P×AInterno LxPxA
- Pharmatherm high performance, holding time: 96 hr, weight: 5,5 kg5,2 L396×396×396 mm145×225×160 mm1 * 1 STSFr. 57.50
Controlled room temperatures profiles
- DescrizioneCapacitàEsterno L×P×AInterno LxPxA
- Pharmatherm controlled room temperature, holding time: 96 hr, weight: 7,6 kg6,6 L396×396×396 mm189×189×185 mm1 * 1 STSFr. 66.10
- Pharmatherm controlled room temperature, holding time: 96 hr, weight: 11,4 kg15,2 L460×460×460 mm249×249×245 mm1 * 1 STSFr. 89.20
- Pharmatherm controlled room temperature, holding time: 96 hr, weight: 18,1 kg26,8 L565×565×565 mm374×319×225 mm1 * 1 STSFr. 133.00