Campylo Thioglycollate Medium Base Dehydrated Culture Media
Catalog # HIMEM908-500G
Supplier: HIMEDIA
New Product
CAS Number:
Specifications
- Conf:1
- Descrizione:Campylo thioglycollate medium base, 500 g
Specifications
About this item
For the isolation, maintenance, and transport of cultures of Campylobacter species.
- Microbiological culture media
- 500 g poly bottles