Label Rolls, Core2Write
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
- Always retain critical documentation, pre-printed and hand-written
- Extremely durable
- Abrasion and tear resistant
- Smear resistant
- Can withstand multiple chemicals
- Can withstand a wide temperature range (–70 to 200 ℃)
- Fully customisable by size, color, pre-printing
All roll labels come with a three inch plastic core. Can be applied to all types of surfaces: glass, poly, stainless steel. Can be autoclaved.
: Also available in sheets (sold separately).
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
- Always retain critical documentation, pre-printed and hand-written
- Extremely durable
- Abrasion and tear resistant
- Smear resistant
- Can withstand multiple chemicals
- Can withstand a wide temperature range (–70 to 200 ℃)
- Fully customisable by size, color, pre-printing
All roll labels come with a three inch plastic core. Can be applied to all types of surfaces: glass, poly, stainless steel. Can be autoclaved.
: Also available in sheets (sold separately).
Product Family Options
Non-sterile
- DescrizioneColoreDimensioniConfezione
- Core2Write label, non-sterilebianco50,8×25,4 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 3’760.00
Specifications
Cat. No.VELTC2WR2X1BLK01 - Core2Write label, non-sterilebianco76,2×25,4 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 3’760.00
Specifications
Cat. No.VELTC2WR3X1BLK01 - Core2Write label, non-sterilebianco76,2×38,1 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 3’760.00
Specifications
Cat. No.VELTC2WR3X1.5BLK01 - Core2Write label, non-sterile, irradiatedbianco101,6×50,8 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 3’760.00
Specifications
Cat. No.VELTC2WR4X2BLK01 - Core2Write label, non-sterilebianco101,6×76,2 mm1000 labels/roll, 4 rolls/case1 * 4.000 ST
Specifications
Cat. No.VELTC2WR4X3BLK01
Sterile
- DescrizioneColoreDimensioniConfezione
- Core2Write label, sterile, irradiatedbianco50,8×25,4 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 4’310.00
Specifications
Cat. No.VELTC2WR2X1BLK02 - Core2Write label, sterile, irradiatedbianco76,2×38,1 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 4’310.00
Specifications
Cat. No.VELTC2WR3X1.5BLK02 - Core2Write label, sterile, irradiatedbianco76,2×25,4 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 4’310.00
Specifications
Cat. No.VELTC2WR3X1BLK02 - Core2Write label, sterile, irradiatedbianco101,6×50,8 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 4’310.00
Specifications
Cat. No.VELTC2WR4X2BLK02 - Core2Write label, sterile, irradiatedbianco101,6×76,2 mm1000 labels/roll, 4 rolls/case1 * 4.000 STSFr. 4’310.00
Specifications
Cat. No.VELTC2WR4X3BLK02