Order Entry
Switzerland
ContactUsLinkComponent
Anti-SLC52A3 Rabbit Polyclonal Antibody
Anti-SLC52A3 Rabbit Polyclonal Antibody
Codice catalogo NOVUNBP2-14683
Nome del fornitore:  Novus Biologicals
New Product
Anti-SLC52A3 Rabbit Polyclonal Antibody
Codice catalogo NOVUNBP2-14683
Nome del fornitore:  Novus Biologicals
New Product

Specifiche

  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • Ospite:
    Rabbit
  • Isotipo:
    IgG
  • Reattività:
    Human
  • ID gene:
    SLC52A3
  • Purezza:
    Immunogen affinity purified
  • Tampone di conservazione:
    PBS (pH 7,2) and 40% glycerol
  • Sequenza:
    This antibody was developed against recombinant protein corresponding to amino acids: RQPRCWEASVEDLLNDQVTLHSIRPREENDLGPAGTVDSSQGQGYLEEKA
  • Temperatura di conservazione:
    Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
  • Conf:
    0,1 mL

Specifiche

Informazioni su questo articolo

SLC52A3 polyclonal specifically detects SLC52A3 in human samples. It is validated for immunohistochemistry, immunohistochemistry-paraffin.

Dilutions: Immunohistochemistry 1:200 to 1:500 and Immunohistochemistry-paraffin 1:200 to 1:500
Application notes: For IHC-Paraffin, HIER pH 6 retrieval is recommended
Preservative: 0,02% sodium azide

Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis. Primary antibodies are guaranteed for one year.

Type:
Antigen: SLC52A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: HuSpecies Glossary