Orders LinkContactUsLinkComponent
Mixing Systems, SpinBag
Mixing Systems, SpinBag
Nome del fornitore:   PARKER HANNIFIN
Mixing Systems, SpinBag
Nome del fornitore:  PARKER HANNIFIN
Certificates
2 varianti
Product Details & Documents
Parker's compact SpinBag Mixing System offers the advantages of single-use in a bench top mixing solution.
  • Constructed of fully validated DuraPure C93 film offering added seal strength, low gas permeability and optical clarity which can be heated to 60 °C
  • Stores flat and ideal for preparation of cell culture media, buffers, bioreactor feeds, and suspension/re-suspension applications
  • Easily integrated with filters, sensors and pumping systems
  • Available with standard configurations, or a variety of ports and tubing options in non-irradiated or gamma irradiated
  • Platinum-cured silicone coated magnetic stir bar offers optimal mixing performance and eliminates the need to validate another material
  • Manufactured in an ISO Class 7 cleanroom
Parker's 3D SpinBag bioprocess containers are constructed of durable, particle-free, DuraPure C93 film, each containing a platinum-cured silicone coated magnetic spin bar. When placed on a mixing plate, the spin bar rotates to create a vortex of motion for thorough mixing/stirring of fluid contents. Parker SpinBags are available in a variety of sizes and port configurations to easily integrate within your application.
Product Family Options
Informazioni sul prodotto
  • Disponibilità
    Price
  • PARKPHSBU010LGI
    SpinBag mixing systems, 10 L
    SpinBag mixing systems, 10 L
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 SET
    1 * 1 SET - SFr. 759.00
    SFr. 759.00
    1 * 1 SET
       SFr. 759.00
  • PARKPHSBU050LGI
    SpinBag mixing systems, 50 L
    SpinBag mixing systems, 50 L
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 SET
    1 * 1 SET - SFr. 784.00
    SFr. 784.00
    1 * 1 SET
       SFr. 784.00
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • SpinBag housing only, 50 L, polycarbonate
    SpinBag housing only, 50 L, polycarbonate
    PARKPHSBH050L
    Log in per disponibilità
    1 * 1 SET - SFr. 743.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • SpinBag housing only, 15 L, polycarbonate
    SpinBag housing only, 15 L, polycarbonate
    PARKPHSBH015L
    Log in per disponibilità
    1 * 1 SET - SFr. 621.00