iFluor® 350-Wheat Germ Agglutinin (WGA) Conjugate
Codice catalogo AATB25525
Nome del fornitore: AAT BIOQUEST
New Product
Numero CAS:
Specifiche
- Conf:1 mg
- Descrizione:iFluor® 350-Wheat Germ Agglutinin (WGA) conjugate
Specifiche
Informazioni su questo articolo
iFluor® 350 WGA conjugate, like Alexa Fluor® 350 WGA conjugate, retains its ability to bind to sialic acid and N-acetylglucosaminyl residues, enhancing its utility in fluorescence imaging and analysis for various scientific investigations.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.