Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in DMSO*
AATB17709
New Product
:
- Conf:0,5 mL
- Descrizione:Gelite™ Safe DNA gel stain *GelRed replacement, 10000X in DMSO*
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
- Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
: For research use only.